This item is now discontinued. Please browse similar items
Allsafe MTL (10179) Combination Googles/Face Shield, Clear Anti-Fog
Representative image only.
Allsafe Mfr # 16671 – Item # RK-81657-30
Combination goggles and shield provides face and eye protection without additional head gear
SPECS & DESCRIPTION
Specifications & Description
  • Frame ColorClear
  • Lens ColorClear
  • Lens CoatingAntifog
  • ComplianceANSI Z87.1+ (High Impact); CSA Z94.3, SEI-certified
  • DescriptionCombination Googles/Face Shield, Clear Anti-Fog
Canadian Standards Association
More About this Item
Shield provides both primary and secondary eye protection. Attached MonoGoggle® goggles feature a soft pliable nose piece and flexible frame for a snug, comfortable fit. Lens gives you distortion-free vision. Clear polycarbonate shield protects your face. Shield features FogGard® Plus anti-fog coating. Compliance: ANSI Z87.1 (High impact), CSA Z94.3, SEI certified