This item is now discontinued. Please browse similar items
Alconox Alcotabs 1500 Critical Cleaning Detergent Tablets; 6 x 100 Tablet Bottles/Cs
Representative image only.
Alconox Mfr # 1500 – Item # RK-17778-15
Biodegradable, concentrated, and free of interfering residue
  • Optimize performance and environmental safety
  • Order new bulk packages for added convenience
SPECS & DESCRIPTION
Specifications & Description
  • FormatTablets
  • Dilution6.5 pH
  • ApplicationsLab pipette cleaning
  • DescriptionCritical Cleaning Detergent Tablets; 6 x 100 Tablet Bottles/Cs
Key Features
  • Optimize performance and environmental safety
  • Order new bulk packages for added convenience
More About this Item
These high-foaming cleaning detergents are suitable for manual cleaning applications. Ideal for use on glass, metal, plastic, rubber, and other hard materials in your critical cleaning processes. All Alconox cleaners rinse completely with no interfering residue. Alconox® and Liquinox® cleaners are biodegradable. All cleaner are safe, odorless, and FDA compliant.
Alcotabs is a effervescent detergent tablet for lab pipette cleaning. Maximum cleaning in syphon pipette washers to clean pipettes and tubes. Slow release bubble action and free rinsing produce reliable results and no interfering residues. USDA authorized. pH 6.5
ALCOJET, ALCONOX, LIQUI-NOX, TERG-A-ZYME-Reg TM Alconox Inc.