This item is now discontinued. Please browse similar items
Air Impurities Removal Systems High-efficiency Filter for use with Ductless Air Filter System
Representative image only.
Air Impurities Removal Systems Mfr # F-987-1 – Item # RK-33703-41
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionHigh-efficiency Filter for use with Ductless Air Filter System
More About this Item
Easily install or change filters in just seconds. Filter packages designed to suit your specific working conditions
High-Efficiency Filters captures smoke and dust — ideal if you mix dry chemicals, sand, grind, or do light welding. If you use solvents or paint in the same process, choose a combination high-efficiency filter with refillable 2" thick carbon module. Filter is 95% efficient.
HEPA Filters capture soldering fumes and vapors that contain lead. If you use solvents or paint in the same process, choose a combination HEPA filter with refillable 2" thick carbon module. Filter is 99.97% efficient.
Activated Carbon Filter captures volatile solvents, touch-up paint fumes, and other noxious odors. The primary