This item is now discontinued. Please browse similar items
Air compressor option (Built-In) for BioDiesel GC System 34009-09
Representative image only.
Item # RK-34009-59
Satisfy and perform all of your requirements for ASTM D6584 with this compact, value-priced system
  • Includes powerful PeakSimple™ USB data system — connect GC and your Windows® computer for a complete analytical system
  • Comprehensive display shows the temperature, pressure, and voltage control
  • Ambient to 400°C temperature controllable column oven
  • Electronic Pressure Control (EPC) for all GC system gases, including carrier gas, to ensure user safety
  • Conforms to the specifications of ASTM D6584
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionAir compressor option (Built-In) for BioDiesel GC System 34009-09
Two year warranty
Key Features
  • Includes powerful PeakSimple™ USB data system — connect GC and your Windows® computer for a complete analytical system
  • Comprehensive display shows the temperature, pressure, and voltage control
  • Ambient to 400°C temperature controllable column oven
  • Electronic Pressure Control (EPC) for all GC system gases, including carrier gas, to ensure user safety
  • Conforms to the specifications of ASTM D6584
  • 2 year warranty
More About this Item
This simple-to-use biodiesel GC system is ideal for the quantitative determination of free and total glycerin in B100 methyl esters by gas chromatography. The calibrated results are displayed directly, eliminating off-line calculations.
Both the carrier gas and the Flame Ionization Detector (FID) combustion gas are controlled by programmable Electronic Pressure Controllers (EPCs). EPCs not only provide rock-solid retention time reproducibility, but allow the carrier gas to be pressure ramped (just as the column oven is temperature ramped) from the built-in PeakSimple data system.
The on-column injector is ideal for 1/8" packed and 0.53-mm wide-bore capillary columns, and is suitable for analyses ranging from methane to heavy, high-boiling hydrocarbons (C44+). The column oven accepts column cage diameters up to four inches, is programmable to 400°C, and recycles quickly with its high-speed cool-down fans.
This traditional four-filament TCD can heat up to 527°F (275°C).
The built-in single-channel PeakSimple data system provides
easy and powerful data acquisition, as well as temperature
programming for the column oven. The column oven is
programmable to 572°F (300°C) with unlimited ramps and holds, and is equipped with fast cool-down fans. EPC for the the carrier gas provides rock-solid retention time reproducibility.