This item is now discontinued. Please browse similar items
Cole-Parmer Essentials Brass Vacuum/Pressure Gauge Connector Kit for all Pumps
Representative image only.
Cole-Parmer Essentials Item # RK-07395-20
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionBrass vacuum/pressure gauge connector kit for all pumps
More About this Item
Dual-head pump gives you increased capacity over the single-head AIR CADET pumps. Use the two pump heads independently or connect them in series with 3/8" ID Tygonr vacuum tubing for up to 23" Hg vacuum.
Dual-head pump includes thread tape made of PTFE PTFE resin, four polyethylene 3/8" NPT(M) x 3/8" hose barb adapters, and a 6-ft, three-wire cord; 115 VAC models include a plug.
Contains two PE adapters and one each of bushing, pipe tee, and adapter for pipe mount.