This item is now discontinued. Please browse similar items
AEMC L642 Thermocouple Model
Representative image only.
AEMC Mfr # 2126.08 – Item # RK-26060-13
Accepts two of the most popular thermocouple types: J, K, T, E, R, S, and N
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Number of Channels2
  • Input TypeTemperature (thermocouple)
  • Temperature Resolution (° F)0.1 (<1000), 1 (>1000)
  • Temperature Accuracy (° F)0.1% to 0.2% + 0.6° to 1°
  • Temperature Accuracy (° C)0.1% to 0.2% + 0.6° to 1°
  • Pressure Resolution0.1 (<1000), 1 (>1000)
  • Sampling Rate8 samples taken at storage interval
  • Memory Size240,000 measurements (512 KB). Recorded data is stored in nonvolatile memory and retained even if battery is low or removed.
  • ConnectionsTwo miniature thermocouple connectors
  • Battery Life100 hours to >45 days (dependent on sample rate and recording length)
  • Battery Type2 x AA batteries
  • DescriptionThermocouple Model
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Compact dual-channel Simple Logger® II data loggers offer programmable storage rates from once every 5 seconds to once per day. Store up to 240,000 readings in the nonvolatile memory. User-selectable storage modes include start/stop, FIFO, and extended recording mode (XRM™). Both models feature USB 2.0 optically isolated communication and include software for data storage, real-time display, analysis, and report generation.
Simple Logger II Thermocouple
Model L642
  • 2 input channels
  • User-selectable thermocouple types: J, K, T, N, E, R, and S
  • Programmable storage rates from 1 per 5 seconds to 1 per day
  • 3 user-selectable storage modes
  • Stores up to 240,000 measurements in nonvolatile memory
  • 5 LED indicators quickly and clearly display logger status
  • CAT III-50 V