This item is now discontinued. Please browse similar items
AEMC 8505 Ground Resistance and Soil Resistivity Tester
Representative image only.
AEMC Mfr # 2136.51 – Item # RK-20025-78
Simple and fast check for transformer coil continuity or capacitor and switch functionality
  • Single-button operation allows rapid inspection of devices, increasing productivity
  • Bright LED indicators and buzzer alert you to status of transformer or capacitor
  • Multifrequency source signals improve test reliability and accuracy
  • Automatically detects device as transformer or capacitor
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • PowerFour AA alkaline batteries (included), 2500 tests typical
  • Width (in)3.625
  • Length (in)7.25
  • Height (in)1.25
  • DescriptionGround Resistance and Soil Resistivity Tester
  • Warranty1 Year
1 Year Warranty
Key Features
  • Single-button operation allows rapid inspection of devices, increasing productivity
  • Bright LED indicators and buzzer alert you to status of transformer or capacitor
  • Multifrequency source signals improve test reliability and accuracy
  • Automatically detects device as transformer or capacitor
  • 1 year warranty
More About this Item
The Quick Tester transformer ratiometer is ideally suited to shipment inspections for faulty units and verification of operational status of installed devices. Use the Quick Tester handheld meter to determine if more in-depth tests should be performed or if repairs are needed. Microprocessor-based design provides control, stability, and repeatability for routine tests.
Detects shorts (<20 ), transformer function (>1 mh), and capacitor function (0.5 µf to <1 mf).