This item is now discontinued. Please browse similar items
Advantec Sterile two-piece monitor, gridded surface, 0.45 um, 50/pack
Representative image only.
Advantec Mfr # 37AS245PS – Item # RK-02935-20
Ideal for bacterial, yeast, mold and other contamination analysis
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • ColorBlack
  • Port Membrane Diameter (µm)0.45
  • Surface MembraneGridded
  • DescriptionSterile two-piece monitor, gridded surface, 0.45 um, 50/pack
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Pre-assembled 2-piece units contain a built-in membrane filter and a support pad. Monitors are suitable for field collection and transportation of samples as well as analysis in the laboratory. Samples collected on the press-fit membrane can then be cultured in place or removed for further analysis. Individually packaged sets are pre-sterilized.