Advantec Aborbent Pads for Petri Dishes, Sterile, Cellulose, 47 mm; 100/Bx

Representative image only.
Advantec Mfr # B200G047A – Item # RK-14005-22
Ideal for use in culturing microorganisms
  • Tested for absorption of 1.8 to 2.2 mL of Culture media
  • pH neutral
  • Individually packaged to assure sterility
  • 1 year warranty

$78.83CAD / Box of 100
Often Bought With…

SPECS & DESCRIPTION

Specifications & Description

  • MaterialCellulose
  • Diameter (mm)47
  • Thickness (mm)0.85
  • Max Absorption1.8 to 2.2 mL
  • DescriptionAborbent Pads for Petri Dishes, Sterile, Cellulose, 47 mm; 100/Bx
1 Year Warranty

Key Features

  • Tested for absorption of 1.8 to 2.2 mL of Culture media
  • pH neutral
  • Individually packaged to assure sterility
  • 1 year warranty

More About this Item

Absorbent pads are white cellulose discs that fit into standard petri dishes. Pre-sterilized and tested for absorption of 1.8 to 2.2 mL of culture media, these pads are ideal for culturing using broth media in petri dishes.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat