This item is now discontinued. Please browse similar items
Advantec GF75293MM Grade GF75 Glass Fiber Filters; 293mm dia; 50/box
Representative image only.
Advantec Mfr # GF75293MM – Item # RK-06656-25
For analytical or gravimetric studies
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialGlass Microfiber
  • Pore Size (micron)0.3
  • Diameter (mm)293
  • Diameter (cm)29.3
  • DescriptionGrade GF75 Glass Fiber Filters; 293mm dia; 50/box
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Highest retention; filtering very fine protein precipitates and clarifying chemically aggressive solutions.