This item is now discontinued. Please browse similar items

Advantec A065F047A Sterile MCE Gridded Filters w/Pads, 0.65um, 47mm Dia; 100/Pk

Representative image only.
Advantec Mfr # A065F047A – Item # RK-06644-43
Presterilized membranes in convenient single-, multi-, or continuous-feed packs
  • Unique high porosity provides superior flow rate Can be safely exposed to temperatures up to 130°C (266°F)
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • MaterialMixed Cellulose Ester
  • Pore Size (µm)0.65
  • Diameter (mm)47
  • SurfaceGridded
  • Absorbent PadIncluded
  • DescriptionSterile MCE Gridded Filters w/Pads, 0.65um, 47mm Dia; 100/Pk
  • Warranty1 year
1 Year Warranty

Key Features

  • Unique high porosity provides superior flow rate Can be safely exposed to temperatures up to 130°C (266°F)
  • 1 year warranty

More About this Item

Mixed cellulose ester membranes are suitable for microbiological analysis of water, wastewater, pharmaceuticals, or beverages. Offered with or without grid lines; grid lines facilitate counting colonies on the membrane surface. Grid squares measure 3.1 mm and represent 1/100 of the area of a 47-mm membrane. Black membranes maximize contrast between colonies and the filter; green membrane enables viewing of black, white, and colorless colonies on a single filter. Presterilized by EtO, these 47-mm diameter filters are available in envelopes of 10, individually packaged, or now individually wrapped continuous feed. Order membranes with absorbent pad for use with broth media.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat