This item is now discontinued. Please browse similar items
Advantec A045A154D Nitrocellulose membranes 0.45um 15x15cm 10/bx
Representative image only.
Advantec Mfr # A045A154D – Item # RK-06645-81
Free of acetate and other esters
  • Maximum protein/nucleic acid binding
  • Low background
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialNitrocellulose
  • Pore Size (micron)0.45
  • Sheet Length (in)6
  • Sheet Width (in)6
  • DescriptionNitrocellulose membranes 0.45um 15x15cm 10/bx
  • Warranty1 year
1 Year Warranty
Key Features
  • Maximum protein/nucleic acid binding
  • Low background
  • 1 year warranty
More About this Item
Available in two pore sizes—0.45 µm is suitable for most blotting applications, 0.20 µm sizes for lower molecular weights.