Advanced Green 3 Low-Bleed Septa, 9 mm AG3 CenterGuide; 50/Pk
Representative image only.
Advanced Green 3 Mfr # 246713 – Item # RK-01999-20
True long-life, high temperature green spectrum
  • Reduced injection port sticking and get more injections per septum
  • 1 month warranty
PRODUCT OPTIONS
$160.60CAD / Pkg of 50
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionSepta, CenterGuide, 9 mm; 50/Pack
  • Warranty1 month
1 Month Warranty
Key Features
  • Reduced injection port sticking and get more injections per septum
  • 1 month warranty
More About this Item
Advanced Green 3 are low-bleed septa that are specifically created to combine a long injection life with low injection port adhesion to meet even your most challenging analysis. Maximum temperature 305°C. Pre-piercing eliminates injection coring on select CenterGuide septa.