This item is now discontinued. Please browse similar items
Advanced Green 3 Low-Bleed Septa, AG3 for Shimadzu; 50/Pk
Representative image only.
Advanced Green 3 Mfr # 246424 – Item # RK-01999-24
True long-life, high temperature green spectrum
  • Reduced injection port sticking and get more injections per septum
  • 1 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionSepta, Shimadzu; 50/Pack
  • Warranty1 month
1 Month Warranty
Key Features
  • Reduced injection port sticking and get more injections per septum
  • 1 month warranty
More About this Item
Advanced Green 3 are low-bleed septa that are specifically created to combine a long injection life with low injection port adhesion to meet even your most challenging analysis. Maximum temperature 305°C. Pre-piercing eliminates injection coring on select CenterGuide septa.