This item is now discontinued. Please browse similar items
Additel 9510 Cal Task Management Software for multifunction calibrator
Representative image only.
Additel Mfr # 9510 – Item # RK-16106-82
A wide array of Additel accessories to support your Additel instruments
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithADT22XA series multifunction process calibrators and ADT761 series automated pressure calibrators
  • DescriptionCal Task Management Software for multifunction calibrator
More About this Item
Choose from a wide range of accessories for your Additel meters including replacement batteries, software, and carrying cases.
The Additel 9510 Additel/Cal was designed specifically for task management of temperature and pressure calibration. The main features include establishing a database of all reference and test instruments, maintaining a customer database, creating, uploading and downloading calibration tasks, generating a calibration report, and customizing the calibration certificate.