This item is now discontinued. Please browse similar items
Accuform tak635 Loop and Lock Tags - Do Not Operate 10/pk
Representative image only.
Accuform Mfr # TAK635 – Item # RK-40504-47
Secure tags in place to warn of hazardous situations
  • Self-locking ribbed plastic tie strap is attached with reinforced metal grommet
  • 8" long strap adjusts and locks for tight security with a 50 lb. pull strength
  • Strap can only be cut for removal
  • Reinforced metal grommet has a 3/8" hole
  • Tag is 15mil thick and measures 5-7/8"H x 3-3/8"W
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionLoop and Lock Tags - Do Not Operate 10/pk
Key Features
  • Self-locking ribbed plastic tie strap is attached with reinforced metal grommet
  • 8" long strap adjusts and locks for tight security with a 50 lb. pull strength
  • Strap can only be cut for removal
  • Reinforced metal grommet has a 3/8" hole
  • Tag is 15mil thick and measures 5-7/8"H x 3-3/8"W