This item is now discontinued. Please browse similar items
Accuform MGNF984VP Sign, Safety First-Report All Accidents No Matter…, 7x10", Plastic; 1/Pk
Representative image only.
Accuform Mfr # MGNF984VP – Item # RK-60710-77
SPECS & DESCRIPTION
Specifications & Description
  • HeaderSAFETY FIRST
  • LanguagesEnglish
  • MaterialPlastic
  • Width (in)10
  • Height (in)7
  • DescriptionSign, Safety First-Report All Accidents No Matter…, 7x10\", Plastic; 1/Pk
More About this Item
These signs are intended to indicate potential, type, and degree of the hazard that may lead to accidental injury or property damage. They are intended to define specific hazards by conveying a clear, easily understood message. The sign headers, and colors used in accident prevention safety signs are specified by OSHA and ANSI to designate the severity of the hazard. Each color has a specific role.
Signs are available in Adhesive Vinyl (VS), Plastic (VP), and Aluminum (VA) to best suit your application. VS is a flexible 4-mil pressure sensitive vinyl with adhesive back and peel-off liner. This material is ideal for low cost, light duty applications. VP is .055” polyethylene plastic and is a good medium duty indoor and sheltered outdoor environments. VA is .040” aluminum with over laminate