Accuform MCHL125VP Safety Sign, Danger - Hot (with icon), 10" x 14", Plastic

Representative image only.
Accuform Mfr # MCHL125VP – Item # RK-40711-85
Protect your workers by clearly displaying possible hot areas

PRODUCT OPTIONS

$23.13CAD / Each

SPECS & DESCRIPTION

Specifications & Description

  • HeaderDANGER
  • LanguagesEnglish
  • MaterialPlastic
  • Width (in)14
  • Height (in)10
  • DescriptionSafety Sign, Danger - Hot (with icon), 10" x 14", Plastic

More About this Item

These signs are intended to indicate potential, type, and degree of the hazard that may lead to accidental injury or property damage. Signs define specific hazards by conveying a clear, easily understood message. The sign headers and colors used in accident prevention safety signs are specified by OSHA and ANSI to designate the severity of the hazard. Each color has a specific role.
DANGER (red): indicates an imminently hazardous situation which, if not avoided, will result in death or serious injury.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat