This item is now discontinued. Please browse similar items
Accuform LZH619PS5 GHS Label, Health Hazard Pictogram, 2" x 2", adhesive paper, 500/roll
Representative image only.
Accuform Mfr # LZH619PS5 – Item # RK-50713-21
SPECS & DESCRIPTION
Specifications & Description
  • HeaderHEALTH HAZARD
  • LanguagesEnglish
  • MaterialAdhesive Paper
  • Width (in)2
  • Height (in)2
  • Quantity500/roll
  • DescriptionGHS Label, Health Hazard Pictogram, 2\" x 2\", adhesive paper, 500/roll
More About this Item
GHS (Globally Harmonized System) is a worldwide program for classifying chemicals in the workplace. GHS recognizes nine hazard pictograms. The pictograms identify the hazard which helps users take proper precautions to minimize adverse effects.
GHS Health Hazard pictogram indicates the following potential hazard: carcinogen, mutagenicity, reproductive toxicity, respiratory sensitizer, target organ toxicity, aspiration toxicity.