This item is now discontinued. Please browse similar items
Accuform LZH615EV2 GHS Label, Gas Cylinder Pictogram, 2" x 2", adhesive poly, 250/roll
Representative image only.
Accuform Mfr # LZH615EV2 – Item # RK-50712-86
SPECS & DESCRIPTION
Specifications & Description
  • HeaderGAS CYLINDER
  • LanguagesEnglish
  • MaterialAdhesive Poly
  • Width (in)2
  • Height (in)2
  • Quantity250/roll
  • DescriptionGHS Label, Gas Cylinder Pictogram, 2\" x 2\", adhesive poly, 250/roll
More About this Item
GHS (Globally Harmonized System) is a worldwide program for classifying chemicals in the workplace. GHS recognizes nine hazard pictograms. The pictograms identify the hazard which helps users take proper precautions to minimize adverse effects.
GHS Gas Cylinder pictogram indicates the following potential hazard: gases under pressure.