This item is now discontinued. Please browse similar items
Accuform LZH613EV2 GHS Label,Exploding Bomb Pictogram,2" x 2",adhesive poly,250/roll
Representative image only.
Accuform Mfr # LZH613EV2 – Item # RK-50712-70
SPECS & DESCRIPTION
Specifications & Description
  • HeaderEXPLODING BOMB
  • LanguagesEnglish
  • MaterialAdhesive Poly
  • Width (in)2
  • Height (in)2
  • Quantity250/roll
  • DescriptionGHS Label,Exploding Bomb Pictogram,2\" x 2\",adhesive poly,250/roll
More About this Item
GHS (Globally Harmonized System) is a worldwide program for classifying chemicals in the workplace. GHS recognizes nine hazard pictograms. The pictograms identify the hazard which helps users take proper precautions to minimize adverse effects.
GHS Exploding Bomb pictogram indicates the following potential hazard: explosives, self-reactives, and organic peroxides.