This item is now discontinued. Please browse similar items
Accuform LZH612PS5 GHS Label,Flame Over Circle Pictogram,2" x 2",adhesive paper,500/roll
Representative image only.
Accuform Mfr # LZH612PS5 – Item # RK-50712-61
SPECS & DESCRIPTION
Specifications & Description
  • HeaderFLAME
  • LanguagesEnglish
  • MaterialAdhesive Paper
  • Width (in)2
  • Height (in)2
  • Quantity500/roll
  • DescriptionGHS Label,Flame Over Circle Pictogram,2\" x 2\",adhesive paper,500/roll
More About this Item
GHS (Globally Harmonized System) is a worldwide program for classifying chemicals in the workplace. GHS recognizes nine hazard pictograms. The pictograms identify the hazard which helps users take proper precautions to minimize adverse effects.
GHS Flame Over Circle pictogram indicates the following potential hazard: oxidizers