This item is now discontinued. Please browse similar items
Accuform LZH612EV5 GHS Label,Flame Over Circle Pictogram,2" x 2",adhesive poly,500/roll
Representative image only.
Accuform Mfr # LZH612EV5 – Item # RK-50712-63
SPECS & DESCRIPTION
Specifications & Description
  • HeaderFLAME
  • LanguagesEnglish
  • MaterialAdhesive Poly
  • Width (in)2
  • Height (in)2
  • Quantity500/roll
  • DescriptionGHS Label,Flame Over Circle Pictogram,2\" x 2\",adhesive poly,500/roll
More About this Item
GHS (Globally Harmonized System) is a worldwide program for classifying chemicals in the workplace. GHS recognizes nine hazard pictograms. The pictograms identify the hazard which helps users take proper precautions to minimize adverse effects.
GHS Flame Over Circle pictogram indicates the following potential hazard: oxidizers