This item is now discontinued. Please browse similar items
Accuform LZH611EV2 GHS Label, Flame Pictogram, 2" x 2", adhesive poly sheet, 250/roll
Representative image only.
Accuform Mfr # LZH611EV2 – Item # RK-50712-54
SPECS & DESCRIPTION
Specifications & Description
  • HeaderFLAME
  • LanguagesEnglish
  • MaterialAdhesive Poly
  • Width (in)2
  • Height (in)2
  • Quantity250/roll
  • DescriptionGHS Label, Flame Pictogram, 2\" x 2\", adhesive poly sheet, 250/roll
More About this Item
GHS (Globally Harmonized System) is a worldwide program for classifying chemicals in the workplace. GHS recognizes nine hazard pictograms. The pictograms identify the hazard which helps users take proper precautions to minimize adverse effects.
GHS Flame pictogram indicates the following potential hazards: flammables, pyrophorics, self-heating, emits flammable gas, self-reactives, and organic peroxides.