This item is now discontinued. Please browse similar items
Accuform KDD134 Universal Toggle/Rocker and Flat Wall Paddle Switch Lockout
Representative image only.
Accuform Mfr # KDD134 – Item # RK-40504-20
Lock out a wall switch in either ''on'' or ''off'' position
  • Install on standard toggle or rocker wall switch using existing wall plate screw
  • To operate, set switch in preferred position, close lockout and secure with padlock to prevent access to the switch
  • Lockout features side-hinge opening
  • Manufactured from visible red plastic
  • Accommodates padlock with shackle diameter up to 9/32'' (7.1 mm)
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionUniversal Toggle/Rocker and Flat Wall Paddle Switch Lockout
Key Features
  • Install on standard toggle or rocker wall switch using existing wall plate screw
  • To operate, set switch in preferred position, close lockout and secure with padlock to prevent access to the switch
  • Lockout features side-hinge opening
  • Manufactured from visible red plastic
  • Accommodates padlock with shackle diameter up to 9/32'' (7.1 mm)