This item is now discontinued. Please browse similar items
Accuform KDD123 Single-Pole No Hole Circuit Breaker Lockout
Representative image only.
Accuform Mfr # KDD123 – Item # RK-40504-06
Lock out electrical power at the source – right at the circuit breaker, no need to lock the entire panel
  • Manufactured of rigid plastic in visible bright red
  • No modifications required, attach directly to the breaker toggle and panel to prevent the breaker from being switched on once padlock is in place
  • Fits over most major brands of breaker toggles, in the off position
  • Manufactured of rigid plastic in visible bright red
  • Accommodates padlocks with shackle diameter up to 9/32" (7.1 mm) (sold separately)
  • Single-pole, no-hole style fits over breaker toggle up to 9/16" (14.3 mm) width without a recessed hole by fastening with a screw (screw included)
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionSingle-Pole No Hole Circuit Breaker Lockout
Key Features
  • Manufactured of rigid plastic in visible bright red
  • No modifications required, attach directly to the breaker toggle and panel to prevent the breaker from being switched on once padlock is in place
  • Fits over most major brands of breaker toggles, in the off position
  • Manufactured of rigid plastic in visible bright red
  • Accommodates padlocks with shackle diameter up to 9/32" (7.1 mm) (sold separately)
  • Single-pole, no-hole style fits over breaker toggle up to 9/16" (14.3 mm) width without a recessed hole by fastening with a screw (screw included)
More About this Item
No need to lock out the entire panel; these compact, universally designed lockouts fit over most major brands of breaker toggles in the "off" position. Accommodates padlocks (not included) with shackle diameter up to 9/32" (7.1 mm).
Single-Pole, No-Hole Style fits over a breaker toggle up to 9/16" (1.4 cm) width without a recessed hole by fastening with a screw (screw included).