96-well Polypropylene Flat Top PCR Microplate, Low Profile, No Skirt, Clear, Nonsterile; 100/Pk
Representative image only.
PCRmax Item # RK-67103-53
Ultra-thin and consistent wall thickness allows precise thermal transfer for optimal results
  • Manufactured with 99.9% pure virgin polypropylene
  • RNAse/DNAse-free, nonpyrogenic, nonsterile
  • Compatible with sealing film
  • Suitable for 0.1 mL thermal cycler blocks
PRODUCT OPTIONS
$658.61CAD / Pkg of 100
SPECS & DESCRIPTION
Specifications & Description
  • Number of Wells96
  • Well Volume (µL)100
  • Well ShapeRound
  • SterileNo
  • Plate TypeNo skirt
  • Plate ColorClear
  • MaterialPolypropylene
  • Description96-well Polypropylene Flat Top PCR Microplate, Low Profile, No Skirt, Clear, Nonsterile; 100/Pk
Key Features
  • Manufactured with 99.9% pure virgin polypropylene
  • RNAse/DNAse-free, nonpyrogenic, nonsterile
  • Compatible with sealing film
  • Suitable for 0.1 mL thermal cycler blocks
More About this Item
PCRmax® microplates are designed to fit real-time PCR thermal cyclers and sequencers. The construction of pure, clear polypropylene allows for full transparency. Microplates are compatible with PCRmax Alpha Cycler instruments.