This item is now discontinued. Please browse similar items
3M Personal Air Monitor, Organic Vapors; 50/Cs
Representative image only.
3M Mfr # 3M-3500B – Item # RK-60060-99
Keep workers safe from hazardous environments
  • Simple and effective — badge-style design ensures easy compliance with air monitoring reulations
SPECS & DESCRIPTION
Specifications & Description
  • Gases MeasuredOrganic Vapor
  • Analysis IncludedNo
  • DescriptionPersonal Air Monitor, Organic Vapors; 50/Cs
Key Features
  • Simple and effective — badge-style design ensures easy compliance with air monitoring reulations
More About this Item
The 3M™ Organic Vapor Passive Air Monitoring badges use a single charcoal sorbent wafer for easy collection of organic vapors with later analysis by gas chromatography. Clip unit to your clothing, wear for the duration of the sampling period, and send it to 3M or any other certified gas chromatography lab to determine exposure.
All 3M Air Monitors are simple and effective devices that collect contaminants through the scientific principle of diffusion. They meet or exceed OSHA accuracy requirements of ±25% at 95% confidence level for many work contaminants.
Choose from standard badges and badges with back-up sections. The Organic Vapor Passive Monitoring Badges with Back-Up Section uses primary and back-up sorbent pads for sampling demanding environments. Especially suited for monitoring compounds like acrylonitrile, butadiene and methylene chloride for which activated carbon shows limited capacity.