This item is now discontinued. Please browse similar items
3M FF-403 Full-Facepiece Respirators, Large
Representative image only.
3M Mfr # FF-403 – Item # RK-40113-22
Lens resists paint and stains for quick and easy cleanup
  • Breath easier with the Cool Flow™ valve
  • Keep the lines of communication open with the passive speaking diaphragm
  • Maximum comfort with a wide field of view
SPECS & DESCRIPTION
Specifications & Description
  • SizeLarge
  • ComplianceNIOSH; ANSI Z87.1-2003
  • ColorBlack
  • DescriptionFull-Facepiece Respirators, Large
Key Features
  • Breath easier with the Cool Flow™ valve
  • Keep the lines of communication open with the passive speaking diaphragm
  • Maximum comfort with a wide field of view
More About this Item
These full-facepiece respirators feature a six-strap design, silicone face seal and nose cup, and comfort cradle that combine for a secure, comfortable fit. Scotchgard™ Protector coating on the lens causes liquids and paint to bead on the surface where they can be easily wiped away. Communicate clearly and easily through the passive speaking diaphragm. Cool Flow valve allows easy breathing, reducing heat and moisture buildup for extra comfort.
Order respirator cartridges separately according to the demands of your work environment.
Compliance: ANSI Z87.1-2003 for high impact
Approval: NIOSH