This item is now discontinued. Please browse similar items
3M 7000 Series Silicone Respirator, Mediuml
Representative image only.
3M Mfr # 7800M – Item # RK-86459-02
Light silicone full facepiece enhances fit and improves durability
  • Wide, scratch-resistant, polycarbonate face shield provides excellent visibility and protection
  • Extra-large inhalation and exhalation valves for easier breathing
SPECS & DESCRIPTION
Specifications & Description
  • SizeMedium
  • ComplianceNIOSH
  • ColorBlack
  • 3M Item Number7800M
  • DescriptionSilicone Respirator, Mediuml
Key Features
  • Wide, scratch-resistant, polycarbonate face shield provides excellent visibility and protection
  • Extra-large inhalation and exhalation valves for easier breathing
More About this Item
Respirator includes a a standard nosecup to prevent fogging and a scratch resistant polycarbonate lens for greater durability. A high quality, rust-free speaking diaphragm facilitates excellent communication. A double flap face seal helps ensure proper fit and protection and four adjustable straps help distribute the respirators weight for better comfort.
Order respirator cartridges separately according to the demands of your work environment. Versatile respirator accepts 6000 series bayonet cartridges.
Approval: NIOSH