This item is now discontinued. Please browse similar items
3M 6900 Full-face respirators, Large
Representative image only.
3M Mfr # 6900 – Item # RK-40113-14
Lightweight and comfortable—ideal for extended wear
  • Exhalation valve directs air downward to prevent fogging of facepiece
SPECS & DESCRIPTION
Specifications & Description
  • SizeLarge
  • ComplianceNIOSH
  • ColorGray
  • DescriptionFull-face respirators, Large
Key Features
  • Exhalation valve directs air downward to prevent fogging of facepiece
More About this Item
Full-facepiece respirators for maximum personal protection. All respirator headstraps are adjustable for a secure fit.
Order respirator cartridges separately according to the demands of your work environment.
Approval: NIOSH
Note: For models 40112-10, -12, and -14, OSHA regulations require gas-proof goggles be worn with half-mask respirators when using formaldehyde.