Product Type
Control Type
Digital Dispensing
Max Flow Rate (LPM)
Max Flow Rate (mL/min)
Max Flow Rate Per Channel (mL/min)
High Flow Rate Peristaltic Pumps
Masterflex Single Use
Custom Disposable Sterile Bioprocess Assemblies.
Custom cut tubing
CUSTOM CUT TUBING
High flow peristaltic pumps combine larger flow capability and flexibility and are ideal for flow cell, sipper, OEM, and larger flow feed and transfer applications. Tubing changes are quick and easy and there are over 20 different tubing formulations. When selecting a pump, choose a system that offers a flow rate that's 50-75% of your systems peak capabilities to get a longer tube life as well as more flexibility.
APPLY FILTERS | 1