Ismatec® Pumps and Pump Tubing
Find a pump for your application
Table Scrolls Horizontally
Pump Multichannel Miniflex Independent Channel Control IPC Digital Piston Gear
Flow Rate/Range - mL/min
  • 0.0002 to 68 (per channel 6 roller)
  • 0.0002 to 57 (per channel 8 roller)
  • 0.0001 to 38 (per channel 12 roller)
  • 0.004 to 365 (w/ single-channel head)
  • 0.004 to 190 (dual-channel head)
0.001 to 43
(per channel depending on drive rpm, rollers, and tubing size)
0.0004 to 44 (depending on tubing size used and model used) 0.025-75 1.7 - 5850
MasterflexLive Enabled Yes Yes No No Yes Yes
# of Channels 2 or 4 1 3 4 to 24 1 1
Pressure Capabilities 35psi 35psi 35psi 35psi 100 psi (6.9 bar) 300psi (20.7bar)
Application Cartridges and stopped tubing ensure standard tension in occlusion bed for repeatable performance in low volume/low flow rate pumping. Twist-and-open design, and customizable pump flow patterns ideal for laboratory use. Control of each channel allows for precise "recipe" filling, i.e., perfusion or media injection. Planetary gears drive rollers in unison along tubing for smoother rotation, greater tube life, and accurate liquid flow for multichannel needs. Inert and accurate with touch-screen programmability for high pressure applications and dispensing sensitive or corrosive liquids. Accurate and pulseless flows for pressurized and differential pressure applications, and filling.
Live Image -
The newest Ismatec Pumps feature our intuitive touchscreen and MasterflexLive
Ismatec Miniflex pump with cloud control
Masterflex Ismatec Reglo cloud-enabled drives feature MasterflexLive® enabling real-time remote pump control and monitoring. Get control of all pump parameters, including speed, flow rate, dispense volume, and more. You can monitor and adjust critical processes running 24/7 without the need for onsite personnel, while push notifications alert you to operating conditions and error messages. Internet connection via Wi-Fi or standard Ethernet (RJ45) jack on rear of drive.
Versatile single-channel & multichannel pumps for accurate low-flow fluid delivery
MasterflexLive Touch Screen continuous modeContinuous Mode
MasterflexLive touch screen time mode
Time Mode
MasterflexLive touch screen volume modeVolume Mode
masterflexlive touch screen remote modeRemote Mode
masterflexlive touch screen calibration modeCalibration Mode
  • Continuous run, volume dispense, & time dispense modes - batch control and interval/off time in volume and time modes
  • Flexible operational control via screen, MasterflexLive, or analogue remote control
  • Network connection via standard Ethernet (RJ45) jack on rear of drive or connectivity via Wi-Fi
  • USB connection allows for downloadable firmware updates to be loaded into the pump
Control and monitor your Reglo pump remotely with MasterflexLive
Intuitive
Intuitive
5" capacitive TFT/RGB touchscreen with a clear, wide viewing angle – adjust parameters with a simple touch.
Versatile
Versatile
Packed with advanced technology, the Reglo can dose & dispense 0.0001 to 365-mL per minute.
Connected
Connected
Controlled via MasterflexLive via WiFi or connected to a stand-alone PC.
Secure
Secure
Safely upload & share data with other users. Data is time stamped, encrypted, stored & backed up 24/7.
compliant
Compliant
Premium MasterflexLive subscription gives you 21 CFR Part 11 & EU Annex 11 compliance.
Complete
Complete
The Reglo arrives as either a complete single or dual channel Miniflex Pump Head, or multichannel cassette head.
Multichannel Pumps
Ismatec Reglo 4-channel 12-roller digital pump
Cassette heads offer space for two or four cartridges for multichannel pumping. Each cartridge accepts 3-stop tubing for easy loading and secure retention. The 6, 8 or 12 rollers are low friction and long wearing for smooth operation and low pulsation.
  • Compact, benchtop pumps optimize ease of use, accuracy, and reliability
  • 5” (12.7 cm) capacitive TFT/RGB touch screen for crisp resolution and a wide viewing angle
  • Flexible operational control via screen, MasterflexLive, or analog remote control
  • Continuous run, volume dispense, and time dispense modes — batch control and interval/off time in volume and time modes.
Miniflex Pumps
Ismatec Miniflex Dual Channel Pump MasterflexLive
Single- or dual-channel pump heads have a wide occlusion angles, preventing fluid backflow. An open-head sensor stops the motor when the pump head opens. Versatile pumps for accurate low-flow fluid delivery – with an intuitive touch-screen interface
  • Compact, benchtop pumps optimize ease of use, accuracy, and reliability
  • 5” (12.7 cm) capacitive TFT/RGB touch screen for crisp resolution and a wide viewing angle
  • Flexible operational control via screen, MasterflexLive, or analog remote control
Piston Pumps
Ismatec Reglo Digital Piston Pump MasterflexLive
Valveless piston pump heads feature small displacement to provide excellent precision. A single moving part means lower maintenance, minimal clogs, and less drift over time—offering millions of cycles of reliable, repeatable pumping. Pump heads are fully adjustable from zero to maximum.
  • Continuous run, volume dispense, and time dispense modes—batch control and interval/off time in volume and time modes
  • Antidrip and speed ramping available in volume dispense mode—ensure accuracy and minimize sample loss
  • Choose a piston pump head to fit your flow needs, or order a convenient system
Gear Pumps
Ismatec Gear Pump Drive
Achieve high pressure and accurate, pulseless flow -- with an intuitive touch-screen interface
  • Compact, benchtop pumps optimize ease of use, accuracy, and reliability
  • Antidrip and speed ramping available in volume dispense mode—ensure accuracy and minimize sample loss
  • Choose a piston pump head to fit your flow needs, or order a convenient system
Ismatec Tubing
Ismatec Pump Tubing
● Over 16 different formulations to meet chemical compatibility for your application
● Designed for optimal operation in Masterflex Ismatec peristaltic pumps
Masterflex Single-Use
The widest selection of Single-Use products.
oem design
Custom product development
Expert peristaltic pump system development.
Ismatec Software Downloads
Download Ismatec software and drivers
Download Ismatec software and drivers
Chemical Compatability
Chemical Compatibility
Need Help with this product?
Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.
Call Us At800-363-5900
Live chat