Product Type
Cap / Stopper Size
Capacity (L)
Capacity (mL)
Capacity (oz)
Graduation Interval (mL)
High Speed (rpm)
Cole-Parmer elements
Cole-Parmer elements
Cole-Parmer® elements™ glass and plasticware are quality products at an price you can afford—perfect for general lab research, sample prep, and analysis needs. The glass is borosilicate glass 3.3; the same glass used in higher-end glassware. The plasticware is chemical resistant polypropylene (PP) and has a temperature tolerance from –15 to 150°C. Our glass selection includes beakers, graduated cylinders, Erlenmeyer flasks, volumetric flasks with glass stoppers, and funnels. The plasticware selection includes beakers, beakers with handle, and graduated cylinders.
APPLY FILTERS
filter byclear all
14 RESULTS
SORT BY
Showing 1 - 14 of 14