Cole-Parmer Technical Library
Cole-Parmer eNEWS January 2009
To view this e-mail as a Web page, copy and paste the following link into your browser’s URL Address window: http://www.coleparmer.com/techinfo/techinfo.asp?referred_id=4053&htmlfile=e-news_FD_01-09.htm
eNEWS you can use
January 2009        
Take our Balance and Scale Survey 
and enter for a chance to win an American Express Gift Card.
   See complete offer details.
   See complete line of balances and scales.
Introduction to Fluorescence
Introduction to Fluorescence
In a time when the price of petroleum is ever-fluctuating and the strenuous quest to find a suitable alternative energy sources is key, some are already using ultrasconics to provide a way to covert vegetable oil and animal fats to biodiesel fuel. Learn more about how this process is maintained in a safe and effective manner with the use of ultrasonic transesterification.
Full Story
Complete selection of Thermo Scientific Quantech™ Fluorometer
Respiratory Protection: Preparing Workplaces for an Influenza Breakout
Respiratory Protection: Preparing Workplaces for an Influenza Breakout
An influenza outbreak in a workplace can deliver a devasting blow to productivity and morale. Learn ways to best protect your place of business from an outbreak and how to maintain operations should a pandemic occur.
Full Story
Selection of ONE-Fit™ Healthcare N95 Particulate Respirators
Tech Challenge
Take the challenge,
see if you’re right...
Long Island Ice Tea Automation
Challenge: A customer wants to automate the production of his Long Island Ice Tea product. The only part he still needs to choose in his production is a valve. He will use gravity feed to add a key ingredient to the batch while monitoring the pH of the mixture. The recipe calls for a precise amount of a particular ingredient be added. The customer would like the valve to be easy to control, in order to ensure too much or too little is not added to each batch. Ideally they would like to control the valve using some sort of controller. Which type of valve should be use to regulate the addition of this key ingredient?
  1. Ball Valve
  2. Direct Lift Solenoid Valve
  3. Proportioning Valve
  4. Solenoid-Operated Pinch Valve
See the answer.
Our technical experts answer questions like this every day. Get the answers to your questions by calling our Application Specialist at 800-323-4340.
Energy Conservation in Industrial Plants
Energy Conservation in Industrial Plants
Conserving energy as a means of saving the environment—and bettering your bottom line—has scarcely been more crucial than today. Find out how with the use of some indispensible tools such as thermal imagers and power quality analyzers you can safely and effectively put an end to unnecessary energy loss in your industrial plant.
Full Article
Complete Selection of Fluke instruments
Cole-Parmer @ your service
New Masterflex L/S Digital Drives
Featured Product!
NEW Masterflex® L/S®
Digital Drives
  • Precision pump drive offers a maintenance-free high-accuracy motor and a new graphical interface for easy setup and operation
  • Brushless motor offers ±0.1% speed control accuracy with a 6000:1 turndown
  • Remote control capabilities include
    scaleable speed control input, start/stop/prime,
    motor direction, and tachometer output
  • Dispense by volume, time, or copy;
    cumulative volume function totalizes
    volume dispensed over multiple batches
  • Flow rates from 0.006 to 3400 mL/min
    with Masterflex L/S pump tubing
  • L/S Variable Speed Digital Drives
  • L/S Computer-Compatible Digital Drives
  • L/S Digital Process Drive
Mecmesin Manual Closure Torque Tester
Featured Product!
Mecmesin
Manual Closure Torque Tester
  • Provides low maintenance and field test capabilities with protective splash-proof housing
  • Low-cost digital torque tester with accuracies of 0.5% full scale deflection
  • Registers torque applied in clockwise
    and counter-clockwise directions,
    allowing you to test loosening and
    tightening of the cap, or closure
  • Fits any shaped sample up to 7-1/2" diameter
Kimble Disposable Serological Pipettes
Featured Product!
Kimble®
Disposable Serological Pipettes
  • Borosilicate glass meets ASTM Specification E438, Type 1, Class B requirements
  • Ideal for tissue culture work
Oakton Waterproof EcoTestr Conductivity and TDS Testers
Featured Product!
Oakton® Waterproof EcoTestr™ Conductivity and TDS Testers
  • Four models to choose from depending on parameter and range requirements
  • Perfect for applications where lost or broken meters are a concern
  • Easy-to-read display
Or visit our
New Product Page
Request your FREE 2009/10 General Catalog today!
  • Visit ColeParmer.com
  • Special Offers!
  • Complete Online Catalog
  • Technical Articles and Buyer’s Guides
  • Request Free Catalogs
  • Give Us Your Opinion
  • Introduction to Fluorescence
  • Respiratory Protection: Preparing Workplaces for an Influenza Breakout
  • Energy Conservation in Industrial Plants
  • CP @ Your Service — Clearance Store
  • Masterflex® L/S® Digital Drives
  • Mecmesin Manual Closure Torque Tester
  • Kimble® Disposable Serological Pipettes
  • Oakton® Waterproof EcoTestr Conductivity and TDS Testers
    Pittcon 2009
    Booth 3855
    March 8-13, 2009
    Ernest N. Morial McCormick Place
    Chicago, IL
    New 2009/10 Cole-Parmer General Catalog
    Spring 2009 Cole-Parmer Preferred Solutions Catalog
    Fluid Handling System Sourcebook
    Plastic Essentials Catalog
    FoodTECH Source 2008
    Canadian customer click here
    Customers in the UK click here
    Customers in China click here
    Customers in India click here
    ©2009 Cole-Parmer. All rights reserved.
    625 East Bunker Ct. Vernon Hills, IL 60061-1844 USA
    This is an advertising message from Cole-Parmer.
    To ensure delivery, add Cole-Parmer to your address book.
    You can modify your profile or unsubscribe anytime.
    Your privacy is important to us—please review our online privacy policy.
    Product Selection Guides  |  Product Maintenance  |  FAQs  |  Technical Library Map
    Articles and White Papers  |  Case Studies  |  Conversion Factors and Tech Data