Cole-Parmer Technical Library
Flowmeter Applications
Example of a chemical proportioning system
using +GF+ Signet Flow Systems
Chemical proportioning system allows for an accurate volumetric ratio to be maintained between the process liquid volume and the chemical injection volume. Real applications include injection or introduction of:
  • vitamins into farm animal drinking water
  • sodium hypochlorite disinfectant into water
  • fertilizer into irrigation water
  • bio-engineered organisms into liquids for manufacturing products such as man-made snow, soaps for large laundries, defoaming chemicals, and insecticides.
Related Links: Printable page E-mail a colleague
Complete Selection of Flowmeters
Flowmeter Applications
Flowmeter FAQs
How Volumetric Flowmeters Work
Installing Your Paddle-Wheel Flow Sensor
Selecting the Right Flowmeter—Part 1
Selecting the Right Flowmeter—Part 2
Product Selection Guides  |  Product Maintenance  |  FAQs  |  Technical Library Map
Articles and White Papers  |  Case Studies  |  Conversion Factors and Tech Data