Chemical Compatibility Chart For Safety Apparel
Average Breakthrough Time (minutes)Average time between contact of chemical on outside of material surface and detection of chemical on inside surface. Industrial chemical breakthrough times are normalized to a permeation rate of 0.1 ug/cm2/min in accordance with ASTM F739-91.
Chemical agents are tested in accordance with Military Standard MIL-STD-282.
Caution: This test data is derived from tests performed on fabric samples only; not finished garments, seams, or component parts.
Compatibility Ratings
| Chemical | Average Breakthrough Time (minutes) | |
| Tyvek QC® | CPF®1 | CPF®2 | CPF®3 | CPF®4 |
| Acetic Acid,100% | NT | NT | >480 | 84 | >480 |
| Acetone,100% | <1 | <4 | 12 | >480 | >480 |
| Acetonitrile, 100% | <1 | <4 | 42 | 7 | >480 |
| Allyl chloride, 99% | NT | NT | 7 | 12 | >480 |
| Benzene, 99% | NT | NT | 36 | >480 | >480 |
| Carbon disulfide, 98% | <1 | <4 | <4 | 16 | >480 |
| Chlorine gas, 100% | NT | NT | >480 | >480 | NT |
| Chlorine liquid, 99% | NT | NT | NT | >480 | >480 |
| Chloroacetic acid, 70-80% | NT | NT | NT | NT | NT |
| Chromic acid, Sat. | NT | NT | >480 | NT | NT |
| Cresols, 100% | NT | NT | >480 | NT | NT |
| Diazinon, 25% | NT | NT | >480 | NT | NT |
| Dichloromethane, 100% | <1 | <4 | <4 | 7 | 114 |
| Dichlorotoluene, mixture | NT | NT | NT | >480 | NT |
| Diethylamine, 98% | <1 | <4 | 14 | >480 | >480 |
| Dimethylformamide, 100% | 7 | 25 | 95 | >480 | >480 |
| Epichlorohydrin, 99% | NT | NT | NT | 67 | >480 |
| Ethyl acetate, 100% | <1 | <4 | 28 | >480 | >480 |
| Ethyl alcohol, 100% | NT | NT | NT | >480 | >480 |
| Ethylene glycol, 99% | NT | >480 | NT | NT | NT |
| Ethylene oxide gas, 100% | NT | NT | NT | >480 | 305 |
| Formaldehyde, 37% | NT | NT | >480 | >480 | >480 |
| Formic acid, 96% | NT | NT | >480 | NT | NT |
| Glutaraldehyde, 50% | NT | >480 | >480 | NT | NT |
| Hexane, 95% | <1 | <4 | 10 | >480 | >480 |
| Hydrazine, 98% | NT | NT | 437 | NT | >480 |
| Hydrochloric acid, 37% | NT | 90 | >480 | >480 | >480 |
| Hydrofluoric acid, 48% | NT | >480 | >480 | 180 | >480 |
| Hydrogen cyanide, 98% | NT | NT | NT | 30 | >480 |
| Hydrogen peroxide, 30% | NT | NT | NT | >480 | NT |
| Isoamyl alcohol, 99% | NT | NT | >480 | NT | NT |
| Jet fuel A, mixture | NT | NT | >480 | NT | NT |
| Malathion, 50% | NT | NT | >480 | NT | NT |
| Methanol, 100% | <1 | 5 | >480 | 8 | >480 |
| Methyl ethyl ketone, 99% | NT | NT | 10 | >480 | >480 |
| Nitric acid, 70% | NT | >480 | >480 | NT | >480 |
| Nitrobenzene, 99% | <1 | 5 | 205 | >480 | >480 |
| Oleum, 27-33 free SO3 | NT | NT | 450 | NT | NT |
| Oleum, 65 free SO3 | NT | NT | NT | 15 | >480 |
| PCB, 50% | NT | NT | >480 | >480 | >480 |
| Phenol, 85% | NT | NT | >480 | 85 | >480 |
| Phosphoric acid, 85% | NT | >480 | >480 | NT | NT |
| Sodium cyanide, 95% | NT | NT | NT | 15 | NT |
| Sodium hydroxide, 50% | >480 | >480 | >480 | >480 | >480 |
| Sodium hypochlorite, 5.25% | NT | NT | >480 | >480 | NT |
| Stoddard solvent, 99% | NT | NT | NT | NT | >480 |
| Styrene, 99% | NT | NT | 12 | >480 | >480 |
| Sulfuric acid, 96% | >480 | >480 | >480 | >480 | >480 |
| Tetrachloroethylene, 100% | <1 | <4 | 10 | >480 | >480 |
| Tetrahydrofuran, 100% | <1 | <4 | <4 | >480 | >480 |
| Titanium tetrachloride, 99% | NT | NT | 15 | 120 | NT |
| Toluene, 100% | <1 | 8 | 4 | >480 | >480 |
| Trichloroethane, 99% | NT | NT | NT | >480 | >480 |
| Vinyl chloride, 100% | NT | NT | >480 | >480 | NT |
| Xylene O, 99% | NT | NT | NT | >480 | >480 |
A Warning: The information in this chart was supplied to Cole-Parmer by other reputable sources and is to be used ONLY as a guide in selecting gloves for chemical compatibility testing. Variability in material thickness, chemical concentration, temperature, and length of exposure to chemicals will affect specific performance.
Always test the products with the specific chemicals and under the specific conditions of your application. Cole-Parmer does not warrant (neither expressed nor implied) the accuracy or completeness of this chart or that any material is suitable for any purpose.
Complete Selection of Safety Products
Safety Apparel Material Selection Guide