YSI 5565 PH/Orp Probe for Multiparameter Meters
Representative image only.
YSI 5565 PH/Orp Probe for Multiparameter Meters
YSI | Mfr: 005565| Item: 9985252
Optimize the performance of YSI meters
$826.57CAD / Each
$826.57CAD / Each
Buy It Together
Current Item
YSI 5565 PH/Orp Probe for Multiparameter Meters
YSI 5565 PH/Orp Probe for Multiparameter Meters
9985252
$826.57CAD / Each
+
YSI Pro30™ Conductivity Meter
5935205
$1,240.59CAD / Each
+ +
YSI Pro1030 Waterproof Handheld pH/Conductivity Meter
5935105
$1,722.39CAD / Each
+
YSI PRO 10 Pro 10 pH/ORP/temperature portable meter
5935216
$1,109.46CAD / Each
+ + +
YSI Pro 1020 pH and Dissolved Oxygen Meter
5935210
$1,720.92CAD / Each
+
YSI 6050020 Pro20 Dissolved Oxygen Meter, waterproof
5935204
$1,196.39CAD / Each
+ =
Total Price 11733.18 ADD 10 ITEMS TO CART
Product Description
Key Features
  • Choose from a variety of accessories
  • Glass-body pH sensor with platium ORP sensor
  • 1 year warranty
More About this Item
Replacement electrodes, cables, power adapters, and other accessories for your YSI water analysis meters.
This probe features a glass body pH sensor and platinum button ORP sensor. The open port of the probe body accomodates this sensor and protects it from damage.
1 Year Warranty
Specifications
  • Compatible WithYSI 556 multiparameter meter
  • DescriptionPH/Orp Probe for Multiparameter Meters