Thermo Scientific Nunc 267245 Microplates, 96-Well, Polypropylene, Non-Treated, Non-Sterile, Round, 0.5 mL; 120/Cs

Representative image only.

Thermo Scientific Nunc 267245 Microplates, 96-Well, Polypropylene, Non-Treated, Non-Sterile, Round, 0.5 mL; 120/Cs

Thermo Scientific | Mfr: 267245| Item: 0192932

Low binding polypropylene is ideal for homogeneous assays and storage

$851.62CAD / Case of 120

$851.62CAD / Case of 120

Buy It Together

Current Item

Thermo Scientific Nunc 267245 Microplates, 96-Well, Polypropylene, Non-Treated, Non-Sterile, Round, 0.5 mL; 120/Cs
Thermo Scientific Nunc 267245 Microplates, 96-Well, Polypropylene, Non-Treated, Non-Sterile, Round, 0.5 mL; 120/Cs
0192932

$851.62CAD / Case of 120
+ =
Total Price 1199.34 ADD 2 ITEMS TO CART
Product Description

Key Features

  • Shared wall design increases the well volume
  • Nontreated polypropylene plates are solvent resistant and autoclavable
  • 1 year warranty

More About this Item

Achieve complete sample recovery with the Thermo Scientific™ Nunc™ MicroWell™ 96-well microplates. Plates are ideal for storage as polypropylene has a lower binding capacity so proteins or DNA will not bind. Withstands temperatures from −80° to 121°C (−112° to 250°F).

Choose from conical bottom wells for optimal sample recovery or round bottom wells which are wider for improved mixing.

Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.

1 Year Warranty
Specifications
  • Number of Wells96
  • Well Volume (mL)0.5
  • Max Working Well Volume (mL)0.45
  • Well ShapeRound
  • Surface TreatmentNon-Treated
  • Lid (Included)No
  • SterileNo
  • Plate ColorNatural
  • Min Temperature (° C)-80
  • Max Temperature (° C)121
  • MaterialPolypropylene
  • Width (mm)86
  • Length (mm)128
  • DescriptionMicroplates, 96-Well, Polypropylene, Non-Treated, Non-Sterile, Round, 0.5 mL; 120/Cs
  • Warranty1 year
Reviews
Q&A