Kimberly-Clark Kimtech Kimwipes EX-L Wipes, 4-1/2" X 8-1/2", 280/box, 60 boxes/case
Representative image only.
Kimberly-Clark Kimtech Kimwipes EX-L Wipes, 4-1/2" X 8-1/2", 280/box, 60 boxes/case
Kimberly-Clark | Mfr: 15407948| Item: 3367004
Low-lint critical task wipes are absorbent, soft, and nonabrasive
$651.98CAD / Case of 16800
BUY MORE & SAVE
  • 1-19$651.98 CAD / Case of 16800
  • 20-49$586.78 CAD / Case of 16800
  • 50-99$554.18 CAD / Case of 16800
  • 100+$521.58 CAD / Case of 16800
$651.98CAD / Case of 16800
BUY MORE & SAVE
  • 1-19$651.98 CAD / Case of 16800
  • 20-49$586.78 CAD / Case of 16800
  • 50-99$554.18 CAD / Case of 16800
  • 100+$521.58 CAD / Case of 16800
Buy It Together
Current Item
Kimberly-Clark Kimtech Kimwipes EX-L Wipes, 4-1/2" X 8-1/2", 280/box, 60 boxes/case
Kimberly-Clark Kimtech Kimwipes EX-L Wipes, 4-1/2" X 8-1/2", 280/box, 60 boxes/case
3367004
$651.98CAD / Case of 16800
+ + +
Parafilm PM999 M Wrap, 4" Wide; 250 Ft/Roll
4400246
$123.03CAD / Each
+
Small Kimwipes Wipe Dispenser, PVC, White
6352810
$54.81CAD / Each
+
Large Kimwipes wipe dispenser 15.1875, White
6352820
$111.83CAD / Each
=
Total Price 1118.16 ADD 6 ITEMS TO CART
Product Description
Key Features
  • Single- and two-ply delicate task wipes are made of a finely textured tissue
  • Solvent-resistant Kimtech CL4 wipes are made of a single-ply, nonwoven fabric specially bonded to eliminate surface fibers and trap particles
  • 3 month warranty
More About this Item
Wipes won’t scratch delicate surfaces and they wipe surfaces clean without leaving residue.
Single-ply Kimwipes® EX-L wipes are made of a finely textured tissue. Come in a convenient Pop-Up® box with antistatic Lintguard® polyshield. Wipes are dispensed individually to reduce waste while minimizing lint, static buildup, and contamination.
3 Month Warranty
Specifications
  • Width (in)4 1/2
  • Length (in)8 1/2
  • DescriptionWipes, 4-1/2" X 8-1/2", 280/box, 60 boxes/case