Humboldt H4200F Adapter Foot for use with Soil Penetrometer
Representative image only.
Humboldt H4200F Adapter Foot for use with Soil Penetrometer
Humboldt | Mfr: H-4200F| Item: 9903912
$95.77CAD / Each
$95.77CAD / Each
Buy It Together
Current Item
Humboldt H4200F Adapter Foot for use with Soil Penetrometer
Humboldt H4200F Adapter Foot for use with Soil Penetrometer
9903912
$95.77CAD / Each
+ =
Total Price 281.86 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • 1 year warranty
More About this Item
Foot is recommended when testing extremely low strength cohesive soils. 1" (25mm) diameter foot.
1 Year Warranty
Specifications
  • DescriptionAdapter Foot to Soil Penetrometer, Measures Low-Stength Cohesive Soils