Humboldt H-5600 Accuflame Bunsen Burner, High-Temperature Burner, Lp Gas
Representative image only.
Humboldt H-5600 Accuflame Bunsen Burner, High-Temperature Burner, Lp Gas
Humboldt | Mfr: H-5600| Item: 3613022
$215.48CAD / Each
BUY MORE & SAVE
  • 1-9$215.48 CAD / Each
  • 10-49$193.93 CAD / Each
  • 50-99$183.16 CAD / Each
  • 100+$172.38 CAD / Each
$215.48CAD / Each
BUY MORE & SAVE
  • 1-9$215.48 CAD / Each
  • 10-49$193.93 CAD / Each
  • 50-99$183.16 CAD / Each
  • 100+$172.38 CAD / Each
Buy It Together
Current Item
Humboldt H-5600 Accuflame Bunsen Burner, High-Temperature Burner, Lp Gas
Humboldt H-5600 Accuflame Bunsen Burner, High-Temperature Burner, Lp Gas
3613022
$215.48CAD / Each
+ =
Total Price 324.66 ADD 2 ITEMS TO CART
Product Description
Key Features
  • 3 month warranty
More About this Item
Gas Burner distributes flame through 100 separate openings in stainless steel grid top for uniform heating. The grid diameter is 11⁄2" (3.8 cm). This burner gives twice the heat of a straight tube burner. This burner features a venturi tube for thorough mixing of gas and air, and adjustable gas orifice and air shutter for complete combustion. Adjustable flame size. Rustproof chrome-plated brass body with zinc-aluminum alloy base.
3 Month Warranty
Specifications
  • Gas TypeLiquid Propane (LP)
  • BTU Output (per hour)3200
  • Inlet Tube OD (in)0.4375
  • Height (in)8 1/2
  • DescriptionAccuflame Bunsen Burner, High-Temperature Burner, Lp Gas
  • Warranty3 Months