Hach 143203 Detergents Color Disc Test Kit ( Toluidine Blue-O Chloroform)
Representative image only.
Hach 143203 Detergents Color Disc Test Kit ( Toluidine Blue-O Chloroform)
Hach | Mfr: 143203| Item: 0554592
Color wheel method provides accurate visual comparisons
$1,066.73CAD / Each
$1,066.73CAD / Each
Buy It Together
Current Item
Hach 143203 Detergents Color Disc Test Kit ( Toluidine Blue-O Chloroform)
Hach 143203 Detergents Color Disc Test Kit ( Toluidine Blue-O Chloroform)
0554592
$1,066.73CAD / Each
+ + =
Total Price 2227.98 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Quick and simple water testing results in an easy-to-carry kit
  • 1 year warranty
More About this Item
Color disc test kits have a color wheel to make visual test matches to estimate concentration in water.
ISO9001 Certified Supplier1 Year Warranty
Specifications
  • TestDetergents
  • Measurement Points0 to 1.2 ppm
  • Reagent PlatformToluidine Blue-o Chloroform
  • Tests Per Kit32
  • DescriptionDetergents Color Disc Test Kit ( Toluidine Blue-O Chloroform)