Cole-Parmer L-Shaped Cell Spreaders, Sterile, Polypropylene, Bags of 10; 500/Cs
Representative image only.
Cole-Parmer L-Shaped Cell Spreaders, Sterile, Polypropylene, Bags of 10; 500/Cs
Cole-Parmer | Item: 0439651
Spread bacterial cultures evenly over agar in petri dishes
$84.28CAD / Case of 500
$84.28CAD / Case of 500
Buy It Together
Current Item
Cole-Parmer L-Shaped Cell Spreaders, Sterile, Polypropylene, Bags of 10; 500/Cs
Cole-Parmer L-Shaped Cell Spreaders, Sterile, Polypropylene, Bags of 10; 500/Cs
0439651
$84.28CAD / Case of 500
+ +
Cole-Parmer Autoclavable Test Tube Basket, PP, 6/pk
0672024
$126.42CAD / Pkg of 6
=
Total Price 272.88 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Sterile to prevent contamination risk
  • L-shape allows application over the entire dish or plate surface
  • Curved end protects agar
More About this Item
Apply cultures over an entire petri dish or plate with these sterile polypropylene cell spreaders. This L-shaped tool has a smooth spreading surface with a slight upward return to reduce the risk of damaging the agar.
Manufactured under ISO 13485 and ISO 9001 quality management systems in an ISO Class 8 (formerly Class 100,000) cleanroom environment, these cell spreaders are radiation sterilized (SAL 10-6), DNase/RNase free, nonpyrogenic, and noncytotoxic. Available individually wrapped or bulk packed.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
Sterile
Specifications
  • SterileYes
  • Overall Length (mm)145
  • Overall Length (in)5 45/64
  • ColorBlue
  • MaterialPolypropylene
  • DescriptionCell Spreaders, Sterile, Polypropylene, 50 bags of 10; 500/Cs