Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Nichrome V inoculating loop with handle, calibrated, 1 µL contained volume
Nichrome  |   Item# RK-14203-25
$21.00 USD/EACH
Are you a reseller or buying in bulk? Call us today.
    Each loop is calibrated to APHA standards. Loops are made of 26-gauge wire. Order insulated loop and needle holder for individual loop separately.
    Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
    Contained Volume 1 µL
    Product Type Inoculating loop
    B & S wire gauge 26
    Loop Length 1 3/4"
    Handle length 6" Insulated
    Qty/ea 1
    Brand Nichrome
    Related products
    Qty/Price
    Availability
    at $145.00 USD/BOX of 500
    In stock.
    Technical Resources
    Virtual Catalog (PDFs)
  • Catalog - English
  •  
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456