Learn more:
[-] Show Less
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
RTD Meters
RTD Meters
  • Resistance Temperature Detectors (RTD)
RTD sensors measure temperature by measuring the change in electrical resistance across the metal wire. The resistance value is then measured and interpreted by a RTD thermometer, and displayed for a user to view. While RTD wire can be made of any metal, platinum is the metal of choice due to its excellent repeatability, stability and resistance to corrosion and chemicals.
Temperature Measurement Applications
Measuring and interpreting temperature has many MRO applications. Measurements throughout the plant can be done quickly and easily and without complex analysis. This information can then be stored electronically for later data trend analysis.
An electrical application is plant wiring. Anytime electrical current flows through a cable or connection, electrical resistance generates heat. Increased current flow through a cable generates more heat. If two similar sized cables have different temperatures, then the cable with the higher temperature carries the higher current. Resistance and heat build-up also occurs at loose or defective wiring connections.
Some typical applications for technicians and service personnel are:
Watching bearing temperatures to predict failure
Detecting motor overload
Finding defective transformers
Detecting loose and improper wiring connections
Checking electrical panels for hot spots
Measuring the surface of HVAC ductwork to reveal temperature variations, poor insulation and duct leakage
Diagnosing blown or plugged steam traps from a distance
Evaluating temperature differences between supply/return registers in a HVAC system
Checking for coil sweating on condensate lines in refrigeration systems
Checking the temperature of emergency relief valves
Diagnosing water leaks by measuring wallboard temperatures. Moist spots on walls will experience water evaporation and therefore will measure at a lower temperature
Testing furnace performance by checking the flue and supply grill temperatures
 Product Detail
Sort
 Availability
 Price
RK-35626-20 Oakton<small><sup>&reg;</sup></small> Acorn<small><sup>&#153;</sup></small> Temp 6 RTD thermometer
Oakton Acorn Temp 6 Thermometer, Pt 100 RTD
Min temperature º C 200,Min temperature º F 320,Max temperature º F 1562
RK-35626-20
In stock.
$192.00 USD/EACH
Qty:
RK-95001-32 Extech 4-Channel Datalogging Thermometer with SD Card Memory
Extech SDL200 4-Channel Datalogging Thermometer with SD Card Memory
Min temperature º C -200,Min temperature º F -328,Max temperature º F 2501
RK-95001-32
In stock.
$349.99 USD/EACH
Qty:
Representative photo only Oakton Temp 360 Datalogging Precision RTD Thermometer w boot
Oakton Temp-360 Datalogging Precision RTD Thermometer w/boot
Min temperature º C 201,Min temperature º F 330,Max temperature º F 2210
RK-91426-60
In stock.
$350.00 USD/EACH
Qty:
RK-93202-51 Hart Scientific Handheld Thermometer w/datalogger Model 1524
Fluke Calibration (Hart Scientific) Handheld Thermometer w/datalogger Model 1524
Product Type Multi-Parameter Thermometers,Brand Fluke Calibration (Hart Scientific)
RK-93202-51
Usually ships in 13 days.
$2,080.00 USD/EACH
Qty:
Representative photo only Fluke Calibration Hart Scientific Handheld Thermometer Model 1523
Fluke Calibration (Hart Scientific) Handheld Thermometer Model 1523
Product Type Multi-Parameter Thermometers,Brand Fluke Calibration (Hart Scientific)
RK-93202-50
In stock.
$1,740.00 USD/EACH
Qty:
Representative photo only Fluke Calibration Hart Scientific Thermometer 4 thermistor RTD inputs
Fluke Calibration (Hart Scientific) Thermometer, 4 thermistor/RTD inputs
Min temperature º C 270,Max temperature º C 1900,Display 1.3 in H x 5 in backlit LCD
RK-93202-10
Usually ships in 12 days.
$5,400.00 USD/EACH
Qty:
Representative photo only Oakton Temp 16 Precision RTD Thermometer w boot
Oakton Temp-16 Precision RTD Thermometer w/boot
Min temperature º C 200,Min temperature º F 328,Max temperature º F 1562
RK-91426-20
In stock.
$205.00 USD/EACH
Qty:
RK-37803-92 Digi-Sense Platinum RTD Thermometer
Digi-Sense Calibrated RTD Thermometer, -58 to 752F/-50 to 400C
Product Type RTD Thermometer,Brand Digi-Sense
RK-37803-92
In stock.
$280.00 USD/EACH
Qty:
Representative photo only Testo RTD Thermocouple Thermometer w Datalogger and Wireless Capability
Testo RTD/Thermocouple Thermometer w/Datalogger and Wireless Capability
Min temperature º C 200,Min temperature º F 328,Max temperature º F 2498
RK-86460-16
Available for order.
Need it sooner?
$655.00 USD/EACH
Qty:
RK-93202-00 Hart Scientific Thermometer, 2 thermocouple and 2 thermistor/RTD inputs
Fluke Calibration 1529 Precision CHUB 4-Channel Thermometer; 2-PRT, 2 T/C
Min temperature º C 270,Max temperature º C 1900,Display 1.3 in H x 5 in backlit LCD
RK-93202-00
Usually ships in 11 days.
$5,400.00 USD/EACH
Qty:
RK-90111-12 Ebro TFX 410 High-accuracy Platinum RTD Thermometer with remote penetration probe
Ebro IP67 RTD Thermometer w/NIST Cal Cert, Remote Sharp Tip; -58-572F
Min temperature º C 50,Min temperature º F 58,Max temperature º F 572
RK-90111-12
In stock.
$265.00 USD/EACH
Qty:
RK-90026-10 Representative image only. See item description for specific details.
ThermoProbe TL1A Intrinsically Safe Stick Thermometer; 12" Long Stem
Min temperature º C -40,Min temperature º F -40,Max temperature º F 400
RK-90026-10
Usually ships in 8 days.
$515.00 USD/EACH
Qty:
Representative photo only HART 1551A 12 Handheld Thermometer Fixed RTD 50 To 160C 1 4x12
HART 1551A-12 Handheld Thermometer, Fixed RTD, –50 To 160C, 1/4x12
Min temperature º C 50,Min temperature º F 58,Max temperature º F 320
RK-93202-08
Usually ships in 8 days.
$680.00 USD/EACH
Qty:
RK-37804-07 Photo shows standard probe, this model comes with a bullet style probe
Digi-Sense Calibrated Scientific Single-Input RTD Thermometer, bullet probe
Min temperature º C 50,Min temperature º F 58,Max temperature º F 302
RK-37804-07
Available for order.
Need it sooner?
$345.00 USD/EACH
Qty:
RK-90026-12 Representative image only. See item description for specific details.
ThermoProbe TL1W Intrinsically Safe Stick Thermometer; w/Horizontal Display, 12" Long
Min temperature º C -40,Min temperature º F -40,Max temperature º F 400
RK-90026-12
Usually ships in 8 days.
$615.00 USD/EACH
Qty:
63 results (showing 1 - 15 )
Per Page