Need Help ?
Our specialists are available to advise you about the best products.
800-363-5900 Live chat
Mixers and Blenders
Mixers and Blenders
Mixers, blenders, and mixing products are common lab tools that can have many variables to consider. We have overhead mixers and lab blenders wth many options. When selecting an overhead mixer or blender for your application, it's important to know container capacity, liquid viscosity, torque requirements, horsepower requirements, rotational speed (rpm) and diameter of the mixing propeller, duty cycle, and power supply.
NEW Cole-Parmer Digital Batch Mixer