log in or register
2,600 pages
of answers
FREE
2013/14 Cole-Parmer
Sourcebook
Our General Catalog features 60,000+ products. View exclusive, hard-to-find items to everyday basics and the brands you've come to trust.
Request your copy today.
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Furnaces
Furnaces
Cole-Parmer offers a range of laboratory furnaces that are able to cover a range of temperatures up to 1600C and capacities up to 22 litres. Our ranges of lab furnaces are ideal for applications including heat treating, reacting studies, sintering processes and ceramic firing.
Important specifications to consider when searching for a laboratory furnace include the process temperature, control requirements, dimensions of sample and inner dimensions of chamber or tube, and pressure range.
Cole-Parmer offers a choice of laboratory furnaces styles including ashing furnace, box or muffle furnace, tube furnace and vacuum furnaces. View our range of laboratory furnaces below or contact one of our application specialists for free support to help find the right product to best suit your needs.
Laboratory Muffle Furnace
These furnaces integrate traditional and modern material to product a combination of performance and reliability. Applications include use in many research facilities for example by chemists in order to bake the moisture out of a sample to ensure it is completely dry.
Ashing furnace
Ashing is also referred to as charring. It is the process where a sample is heated until the only substance remaining is a non-combustible ash. This ash is then analysed for its elemental composition normally via the technique of graphite furnace atomic absorption spectrometry
Tube furnace
A tube furnace heats a tube, typically 0.5 to 1m in length. Samples are placed in the tube and if required a thermocouple inserted in to the tube. One of the main advantages tube furnaces have over other types of furnaces is the ends of the tubes usually protrude from the end of the furnace. As these ends do not get very hot a variety of adapters can be placed on the ends. This allows reactions to be carried out under controlled atmosphere (eg. Pure oxygen, nitrogen and CO/CO2) by setting up gas flow in one end of the tube.
Additional options on tube furnaces include multi zone, meaning that that the tube furnace has multiple heating elements that can be individually controlled. This is important for techniques such as vapour transport synthesis or chemical vapour deposition. Multi-position furnaces allow the chamber to be angled or slowly rotated which can aide slow cooling or crystal growth.
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
 Product Detail
Sort
 Availability
 Price
Representative photo only Furnaces with three stage programmable control 565 cu in overall dimensions 15 13 16 x 16 x 17 1800 watts 200 250V
Furnaces with three-stage programmable control; 565 cu in; overall dimensions; 15 13/16" x 16" x 17"; 1800 watts; 200-250V
Product Type Muffle Furnaces,Minimum temperature º C 50,Maximum temperature º C 1100
RK-33855-35
Usually ships in 6 days or sooner.
$2,575.00 USD/EACH
Qty:
Representative photo only StableTemp furnace with programmable control 2592 cu in 208 240 VAC
StableTemp furnace with programmable control, 2592 cu in, 208/240 VAC
Product Type Muffle Furnaces,Minimum temperature º C 100,Maximum temperature º C 1100
RK-33858-80
Call 1-888-358-4705 for availability.
$4,145.00 USD/EACH
Qty:
RK-33855-30 Model 33855-10 pictured.  Actual furnace may vary slightly.
RK-33855-30
In stock.
$2,600.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Lindberg Blue M Mini Mite Tube Furnace with 16 segment controller 120 VAC
Thermo Scientific Lindberg Blue M Mini-Mite Tube Furnace with 16-segment controller, 120 VAC
Product Type Tube Furnace,Temperature range (º C) low 100,Temperature range (º C) high 1100
RK-33850-00
Usually ships in 9 days.
$2,700.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Thermolyne Compact Benchtop Muffle Furnace 127 5 cu in 120V
Thermo Scientific Thermolyne Compact Benchtop Muffle Furnace; 127.5 cu in; 120V
Product Type Muffle Furnaces,Minimum temperature º C 100,Maximum temperature º C 1100
RK-33900-10
In stock.
$1,940.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Thermolyne Benchtop Muffle Furnace with 350 cu in chamber 120V
Thermo Scientific Thermolyne Benchtop Muffle Furnace with 350 cu in chamber; 120V
Product Type Muffle Furnaces,Minimum temperature º C 100,Maximum temperature º C 1200
RK-02053-12
Call 1-888-358-4705 for availability.
$3,220.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Lindberg Blue M Mini Mite Tube Furnace with 16 segment controller 208 240 VAC
Thermo Scientific Lindberg Blue M Mini-Mite Tube Furnace with 16-segment controller, 208/240 VAC
Product Type Tube Furnace,Temperature range (º C) low 100,Temperature range (º C) high 1100
RK-33850-05
Call 1-888-358-4705 for availability.
$2,980.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Thermolyne Extra Large Chamber Muffle Furnace 2765 cu in 208 VAC 50 60 Hz
Thermo Scientific Thermolyne Extra Large Chamber Muffle Furnace; 2765 cu in; 208 VAC; 50/60 Hz
Product Type Muffle Furnaces,Maximum temperature º C 1093,Maximum temperature º F 1999
RK-33900-20
Call 1-888-358-4705 for availability.
$11,900.00 USD/EACH
Qty:
RK-33855-00 Furnaces with SSP control; 130 cu in; overall dimensions; 11 1/2
Furnaces with SSP control; 130 cu in; overall dimensions; 11 1/2" x 13 13/16 x 13"; 1400 watts; 100-120V
Product Type Muffle Furnaces,Minimum temperature º C 200,Maximum temperature º C 1100
RK-33855-00
In stock.
$1,355.00 USD/EACH
Qty:
Representative photo only StableTemp furnace with basic control 2592 cu in 208 240 VAC
StableTemp furnace with basic control, 2592 cu in, 208/240 VAC
Product Type Muffle Furnaces,Minimum temperature º C 100,Maximum temperature º C 1100
RK-33858-20
Call 1-888-358-4705 for availability.
$3,410.00 USD/EACH
Qty:
Representative photo only Furnaces with SSP Digital control 550 cu in overall dimensions 15 13 16 x 16 x 17 120V
Furnaces with SSP Digital control; 550 cu in; overall dimensions; 15 13/16" x 16" x 17"; 120V
Product Type Muffle Furnaces,Minimum temperature º C 200,Maximum temperature º C 1100
RK-33855-26
Usually ships in 9 days.
$1,680.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Thermolyne Compact Benchtop Muffle Furnace 76 cu in 120V
Thermo Scientific Thermolyne Compact Benchtop Muffle Furnace; 76 cu in; 120V
Product Type Muffle Furnaces,Minimum temperature º C 100,Maximum temperature º C 1100
RK-33900-00
Call 1-888-358-4705 for availability.
$1,585.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Lindberg Blue M Muffle Furnace 324 cu in 120V
Thermo Scientific Lindberg Blue M Muffle Furnace, 324 cu in; 120V
Product Type Muffle Furnaces,Minimum temperature º C 0,Maximum temperature º C 1100
RK-33852-40
Call 1-888-358-4705 for availability.
$3,550.00 USD/EACH
Qty:
Representative photo only Furnaces with three stage programmable control 130 cu in overall dimensions 11 1 2 x 13 13 16 x 13 1400 watts 100 120V
Furnaces with three-stage programmable control; 130 cu in; overall dimensions; 11 1/2" x 13 13/16 x 13"; 1400 watts; 100-120V
Product Type Muffle Furnaces,Minimum temperature º C 50,Maximum temperature º C 1100
RK-33855-10
Call 1-888-358-4705 for availability.
$2,285.00 USD/EACH
Qty:
Representative photo only Thermo Scientific Thermolyne Benchtop Muffle Furnace with 120 cu in chamber 120V
Thermo Scientific Thermolyne Benchtop Muffle Furnace with 120 cu in chamber; 120V
Product Type Muffle Furnaces,Minimum temperature º C 100,Maximum temperature º C 1200
RK-02053-00
Usually ships in 15 days.
$2,750.00 USD/EACH
Qty:
187 results (showing 1 - 15 )
Per Page